missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitrotoluene, 97%

Catalog No. AAL0085614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0085614 25 g
AAL0085606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0085614 Supplier Thermo Scientific Chemicals Supplier No. L0085614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 606-20-2 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 606-20-2
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00007158
InChI Key XTRDKALNCIHHNI-UHFFFAOYSA-N
Synonym 2,6-dinitrotoluene, 2,6-dnt, benzene, 2-methyl-1,3-dinitro, 1-methyl-2,6-dinitrobenzene, 2,6-dinitromethylbenzene, toluene, 2,6-dinitro, rcra waste number u106, 2-methyl-1,3-dinitro-benzene, unii-gg7fav92mk, ccris 1006
PubChem CID 11813
ChEBI CHEBI:957
IUPAC Name 2-methyl-1,3-dinitrobenzene
SMILES CC1=C(C=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 58°C to 65°C
Boiling Point 285°C
Flash Point 207°C (404°F)
Quantity 25 g
UN Number UN3454
Beilstein 2052046
Formula Weight 182.14
Percent Purity 97%
Chemical Name or Material 2,6-Dinitrotoluene
Hazard Category H301+H311+H331-H341-H350-H361f-H373
Hazard Statement GHS H Statement
H301-H311-H331-H350-H341-H361-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROTOLUENES, SOLID
EINECSNumber 210-106-0
RTECSNumber XT1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only