missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylthiophenol, 97%

Catalog No. AAA1328614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1328614 25 g
AAA1328606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1328614 Supplier Thermo Scientific Chemicals Supplier No. A1328614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-72-9 | C8H9S | 137.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-72-9
Molecular Formula C8H9S
Molecular Weight (g/mol) 137.22
MDL Number MFCD00010021
InChI Key QCLJODDRBGKIRW-UHFFFAOYSA-M
Synonym 2,6-dimethylthiophenol, 2,6-dimethyl thiophenol, benzenethiol, 2,6-dimethyl, 2,6-xylenethiol, 2,6-xylyl mercaptan, 2,6-dimethylphenylthiol, 2,6-dimethyl-benzenethiol, fema no. 3666, 2,6-dimethylbenzolthiol, 2,6-thioxylenol
PubChem CID 61045
IUPAC Name 2,6-dimethylbenzenethiol
SMILES CC1=CC=CC(C)=C1[S-]

Specifications

Density 1.038
Boiling Point 122°C (50 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.5749
Quantity 25 g
UN Number UN2810
Beilstein 1099405
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylthiophenol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 204-272-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only