missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpyrazine 98.0+%, TCI America™

Catalog No. p-7126769
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15275G 5 g
D152725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15275G Supplier TCI America Supplier No. D15275G
Only null left

Chemical Identifiers

CAS 108-50-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006148
InChI Key HJFZAYHYIWGLNL-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dimethyl, 3,5-dimethylpyrazine, 2,6-dimethylpiazine, 2,6-dimethylparadiazine, 2,6-dimethyl-1,4-diazine, ccris 2930, unii-n77q72c9i3, fema no. 3273, natural, 2,6-dimethyl pyrazine
PubChem CID 7938
IUPAC Name 2,6-dimethylpyrazine
SMILES CC1=CN=CC(=N1)C

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 155°C
Quantity 5 g
UN Number 1325
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dimethylpyrazine
EINECSNumber (5)-0950
RTECSNumber UQ2975000
TSCA Yes