accessibility menu, dialog, popup

UserName

CH3NH3CN

CAS RN 108-50-9

IUPAC Name: 2,6-dimethylpyrazine
Synonyms: 2,6-dimethylpyrazine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: HJFZAYHYIWGLNL-UHFFFAOYSA-N
SMILES: CC1=CN=CC(C)=N1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)