missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. d36331g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36331G 1 g
D36335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36331G Supplier TCI America Supplier No. D36331G
Only null left

Chemical Identifiers

CAS 100379-00-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01009693
InChI Key ZXDTWWZIHJEZOG-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za
PubChem CID 583322
IUPAC Name (2,6-dimethylphenyl)boronic acid
SMILES CC1=CC=CC(C)=C1B(O)O

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 149.98
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 2,6-Dimethylphenylboronic Acid (contains varying amounts of Anhydride)