missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylphenyl Isocyanate 98.0+%, TCI America™
$125.13 - $425.46
Chemical Identifiers
| CAS | 28556-81-2 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.18 |
| MDL Number | MFCD00002012 |
| InChI Key | YQLRKXVEALTVCZ-UHFFFAOYSA-N |
| Synonym | 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene |
| PubChem CID | 98787 |
| IUPAC Name | 2-isocyanato-1,3-dimethylbenzene |
| SMILES | CC1=CC=CC(C)=C1N=C=O |
Chemical Identifiers
| 28556-81-2 | |
| 147.18 | |
| YQLRKXVEALTVCZ-UHFFFAOYSA-N | |
| 98787 | |
| CC1=CC=CC(C)=C1N=C=O |
| C9H9NO | |
| MFCD00002012 | |
| 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene | |
| 2-isocyanato-1,3-dimethylbenzene |
Specifications
| 28556-81-2 | |
| 113°C | |
| Clear Liquid at 20°C | |
| MFCD00002012 | |
| 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene | |
| CC1=CC=CC(C)=C1N=C=O | |
| 147.18 | |
| 147.18 | |
| 2,6-Dimethylphenyl Isocyanate |
| Yellow | |
| 5 g | |
| C9H9NO | |
| 2206 | |
| YQLRKXVEALTVCZ-UHFFFAOYSA-N | |
| 2-isocyanato-1,3-dimethylbenzene | |
| 98787 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator