missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl Isocyanate 98.0+%, TCI America™

Catalog No. I04105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04105G 5 g
I041025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04105G Supplier TCI America Supplier No. I04105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28556-81-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00002012
InChI Key YQLRKXVEALTVCZ-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene
PubChem CID 98787
IUPAC Name 2-isocyanato-1,3-dimethylbenzene
SMILES CC1=CC=CC(C)=C1N=C=O

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2206
Boiling Point 113°C
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 2,6-Dimethylphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator