missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isocyanate, 98%

CAS: 28556-81-2 | C9H9NO | 147.18 g/mol

$142.11 - $467.81

Chemical Identifiers

CAS 28556-81-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00002012
InChI Key YQLRKXVEALTVCZ-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene
PubChem CID 98787
IUPAC Name 2-isocyanato-1,3-dimethylbenzene
SMILES CC1=CC=CC(C)=C1N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1280506
View Documents
Thermo Scientific Chemicals
L1280506
5 g
Each for $142.11
Only null left
 
AAL1280514
View Documents
Thermo Scientific Chemicals
L1280514
25 g
Each for $467.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28556-81-2
147.18
YQLRKXVEALTVCZ-UHFFFAOYSA-N
98787
CC1=CC=CC(C)=C1N=C=O
C9H9NO
MFCD00002012
2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene
2-isocyanato-1,3-dimethylbenzene

Specifications

28556-81-2
71°C to 72°C (3 mmHg)
1.5356
MFCD00002012
742593
YQLRKXVEALTVCZ-UHFFFAOYSA-N
2-isocyanato-1,3-dimethylbenzene
98787
98%
1.057
5 g
C9H9NO
UN2206
2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene
CC1=CC=CC(C)=C1N=C=O
147.18
147.18
2,6-Dimethylphenyl isocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P260-P264b-P270-P271-P272-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c

H227-H300+H330-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only