missing translation for 'onlineSavingsMsg'
Learn More

[(2,6-Dimethylphenyl)amino](oxo)acetic Acid 98.0+%, TCI America™

Catalog No. D42651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42651G 1 g
D42655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42651G Supplier TCI America Supplier No. D42651G

Chemical Identifiers

CAS 2903-48-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD10018453
InChI Key JFTAPSBIODPQJN-UHFFFAOYSA-N
Synonym (2,6-Dimethylanilino)(oxo)acetic Acid, N-(2,6-Dimethylphenyl)oxamic Acid, DMPAO
PubChem CID 14987869
IUPAC Name 2-(2,6-dimethylanilino)-2-oxoacetic acid
SMILES CC1=C(C(=CC=C1)C)NC(=O)C(=O)O

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material [(2,6-Dimethylphenyl)amino](oxo)acetic Acid