missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylhydroquinone 98.0+%, TCI America™

Catalog No. p-7127435
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26675G 5 g
D266725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26675G Supplier TCI America Supplier No. D26675G

Chemical Identifiers

CAS 654-42-2
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00016466
InChI Key SGWZVZZVXOJRAQ-UHFFFAOYSA-N
Synonym 2,5-Dihydroxy-m-xylene
PubChem CID 69560
IUPAC Name 2,6-dimethylbenzene-1,4-diol
SMILES CC1=CC(O)=CC(C)=C1O

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 138.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethylhydroquinone
EINECSNumber (4)-0057
RTECSNumber MX5900000
TSCA Yes