missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylcyclohexanol (mixture of isomers) 99.0+%, TCI America™
Supplier: TCI America D157825ML
Specifications
| 2,6-Dimethylcyclohexanol (mixture of isomers) | |
| Colorless | |
| C8H16O | |
| 25 mL | |
| 2,6-dimethylcyclohexanol, cyclohexanol, 2,6-dimethyl, 2,6-dimethylcyclohexyl alcohol, ambsclk-255, cyclohexanol,6-dimethyl, 2,6-dimethyl-cyclohexanol, acmc-20ao81, 2,6-dimethylcyclohexanol,c&t, 2,6-dimethyl-cyclohexan-1-ol, 2,6-dimethylcyclohexanol, mixture of isomers | |
| CC1CCCC(C1O)C | |
| 128.215 | |
| 128.22 | |
| Reagent |
| 5337-72-4 | |
| 173°C | |
| MFCD00001502 | |
| 1987 | |
| MOISVRZIQDQVPF-UHFFFAOYSA-N | |
| 2,6-dimethylcyclohexan-1-ol | |
| 21428 | |
| ≥99.0% (GC) | |
| Liquid |
Chemical Identifiers
| 5337-72-4 | |
| 128.215 | |
| MOISVRZIQDQVPF-UHFFFAOYSA-N | |
| 21428 | |
| CC1CCCC(C1O)C |
| C8H16O | |
| MFCD00001502 | |
| 2,6-dimethylcyclohexanol, cyclohexanol, 2,6-dimethyl, 2,6-dimethylcyclohexyl alcohol, ambsclk-255, cyclohexanol,6-dimethyl, 2,6-dimethyl-cyclohexanol, acmc-20ao81, 2,6-dimethylcyclohexanol,c&t, 2,6-dimethyl-cyclohexan-1-ol, 2,6-dimethylcyclohexanol, mixture of isomers | |
| 2,6-dimethylcyclohexan-1-ol |
Safety and Handling
TSCA : No