accessibility menu, dialog, popup

UserName

CAS RN 5337-72-4

IUPAC Name: 2,6-dimethylcyclohexan-1-ol
Synonyms: 2,6-dimethylcyclohexanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: MOISVRZIQDQVPF-UHFFFAOYNA-N
SMILES: CC1CCCC(C)C1O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)