missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylcyclohexanol, 99%, mixture of isomers

Catalog No. p-116734
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149810050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC149810050 Supplier Thermo Scientific Chemicals Supplier No. 149810050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5337-72-4 | C8H16O | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5337-72-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.21
MDL Number MFCD00001502
InChI Key MOISVRZIQDQVPF-UHFFFAOYSA-N
Synonym 2,6-dimethylcyclohexanol, cyclohexanol, 2,6-dimethyl, 2,6-dimethylcyclohexyl alcohol, ambsclk-255, cyclohexanol,6-dimethyl, 2,6-dimethyl-cyclohexanol, acmc-20ao81, 2,6-dimethylcyclohexanol,c&t, 2,6-dimethyl-cyclohexan-1-ol, 2,6-dimethylcyclohexanol, mixture of isomers
PubChem CID 21428
IUPAC Name 2,6-dimethylcyclohexan-1-ol
SMILES CC1CCCC(C1O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9400g/mL
Boiling Point 174.4°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4585 to 1.4615
Linear Formula (CH3)2C6H9OH
Quantity 5 g
Beilstein 06, 18
Specific Gravity 0.94
Formula Weight 128.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethylcyclohexanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-264-9
TSCA TSCA

RUO – Research Use Only