missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzyl alcohol, 98%

CAS: 62285-58-9 | C9H12O | 136.194 g/mol

Supplier:  Thermo Scientific Chemicals H6140903

 View more versions of this product

Catalog No. AAH6140903

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,6-Dimethylbenzyl alcohol
C9H12O
1 g
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
(2,6-dimethylphenyl)methanol
186748
98%
62285-58-9
MFCD00128004
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl
CC1=C(C(=CC=C1)C)CO
136.194
136.19
Solid

Chemical Identifiers

62285-58-9
136.194
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
186748
CC1=C(C(=CC=C1)C)CO
C9H12O
MFCD00128004
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl
(2,6-dimethylphenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only