missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylbenzothiazole 98.0+%, TCI America™
Supplier: TCI America D367425G
Specifications
| 2,6-Dimethylbenzothiazole | |
| Yellow | |
| C9H9NS | |
| 25 g | |
| CC1=NC2=CC=C(C)C=C2S1 | |
| 163.24 | |
| 163.24 | |
| Liquid |
| 2941-71-1 | |
| 132°C | |
| MFCD00022880 | |
| JEKCSLMWKCKDCC-UHFFFAOYSA-N | |
| 2,6-dimethyl-1,3-benzothiazole | |
| 76253 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2941-71-1 | |
| 163.24 | |
| JEKCSLMWKCKDCC-UHFFFAOYSA-N | |
| 2,6-dimethyl-1,3-benzothiazole |
| C9H9NS | |
| MFCD00022880 | |
| 76253 | |
| CC1=NC2=CC=C(C)C=C2S1 |
Safety and Handling
TSCA : Yes