accessibility menu, dialog, popup

UserName

H3CSCH3N

CAS RN 2941-71-1

IUPAC Name: 2,6-dimethyl-1,3-benzothiazole
Synonyms: 2,6-dimethyl-1,3-benzothiazole
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: JEKCSLMWKCKDCC-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(C)C=C2S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results