missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzothiazole 98.0+%, TCI America™

Catalog No. D36745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36745G 5 g
D367425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36745G Supplier TCI America Supplier No. D36745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2941-71-1
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00022880
InChI Key JEKCSLMWKCKDCC-UHFFFAOYSA-N
PubChem CID 76253
IUPAC Name 2,6-dimethyl-1,3-benzothiazole
SMILES CC1=NC2=CC=C(C)C=C2S1

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 g
Formula Weight 163.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethylbenzothiazole