missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylbenzoic Acid 98.0+%, TCI America™
$98.79 - $385.86
Chemical Identifiers
| CAS | 632-46-2 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.177 |
| MDL Number | MFCD00002483 |
| InChI Key | HCBHQDKBSKYGCK-UHFFFAOYSA-N |
| Synonym | benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid |
| PubChem CID | 12439 |
| ChEBI | CHEBI:64827 |
| IUPAC Name | 2,6-dimethylbenzoic acid |
| SMILES | CC1=C(C(=CC=C1)C)C(=O)O |
Chemical Identifiers
| 632-46-2 | |
| 150.177 | |
| HCBHQDKBSKYGCK-UHFFFAOYSA-N | |
| 12439 | |
| 2,6-dimethylbenzoic acid |
| C9H10O2 | |
| MFCD00002483 | |
| benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid | |
| CHEBI:64827 | |
| CC1=C(C(=CC=C1)C)C(=O)O |
Specifications
| 116°C | |
| White | |
| 5 g | |
| C9H10O2 | |
| benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid | |
| CC1=C(C(=CC=C1)C)C(=O)O | |
| 150.177 | |
| CHEBI:64827 | |
| ≥98.0% (GC,T) |
| 632-46-2 | |
| 160°C | |
| Crystal-Powder at 20°C | |
| MFCD00002483 | |
| HCBHQDKBSKYGCK-UHFFFAOYSA-N | |
| 2,6-dimethylbenzoic acid | |
| 12439 | |
| 150.18 | |
| 2,6-Dimethylbenzoic Acid |
Safety and Handling
RTECSNumber : DG8734010
TSCA : No