missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzoic acid, 98+%

Catalog No. AAA1501406
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1501406 5 g
AAA1501403 1 g
AAA1501414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1501406 Supplier Thermo Scientific Chemicals Supplier No. A1501406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 632-46-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 632-46-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002483
InChI Key HCBHQDKBSKYGCK-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid
PubChem CID 12439
ChEBI CHEBI:64827
IUPAC Name 2,6-dimethylbenzoic acid
SMILES CC1=C(C(=CC=C1)C)C(=O)O

Specifications

Melting Point 114°C to 117°C
Quantity 5 g
Odor Odorless
Beilstein 1863791
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material 2,6-Dimethylbenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-177-0
RTECSNumber DG8734010
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only