missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylbenzeneboronic acid, 97%
Description
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 120°C to 126°C (decomposition) |
| Quantity | 1 g |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 149.99 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,6-Dimethylbenzeneboronic acid |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only