missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzaldehyde 97.0+%, TCI America™

Catalog No. p-7127861
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36815G 5 g
D368125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36815G Supplier TCI America Supplier No. D36815G
Only null left

Chemical Identifiers

CAS 1123-56-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00128003
InChI Key QOJQBWSZHCKOLL-UHFFFAOYSA-N
Synonym m-xylene-2-carboxaldehyde, 2,6-dimethyl-benzaldehyde, 2-formyl-m-xylene, benzaldehyde, 2,6-dimethyl, 2,6-dimethylbenzaldehyd, acmc-1bown, 2,6-dimethyl benzaldehyde, ksc493s1p, pharmabridge p-2713, 2,6-dimethylbenzaldehyde
PubChem CID 583841
IUPAC Name 2,6-dimethylbenzaldehyde
SMILES CC1=C(C(=CC=C1)C)C=O

Specifications

Color White-Yellow
Boiling Point 230°C
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dimethylbenzaldehyde