missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzaldehyde, 97%

Catalog No. AAH6450306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6450306 5 g
AAH6450314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6450306 Supplier Thermo Scientific Chemicals Supplier No. H6450306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1123-56-4 | C9H10O | 134.178 g/mol

An aldehyde containing building block used for proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An aldehyde containing building block used for proteomics research

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1123-56-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00128003
InChI Key QOJQBWSZHCKOLL-UHFFFAOYSA-N
Synonym m-xylene-2-carboxaldehyde, 2,6-dimethyl-benzaldehyde, 2-formyl-m-xylene, benzaldehyde, 2,6-dimethyl, 2,6-dimethylbenzaldehyd, acmc-1bown, 2,6-dimethyl benzaldehyde, ksc493s1p, pharmabridge p-2713, 2,6-dimethylbenzaldehyde
PubChem CID 583841
IUPAC Name 2,6-dimethylbenzaldehyde
SMILES CC1=C(C(=CC=C1)C)C=O

Specifications

Melting Point 27°C to 30°C (literature)
Color White to Yellow
Boiling Point 230°C (literature)
Flash Point 96°C (205°F)
Refractive Index 1.548
Quantity 5 g
Beilstein 907019
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 134.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,6-Dimethylbenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only