missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylanisole, 98+%

Catalog No. aab2036514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2036514 25 g
AAB2036522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2036514 Supplier Thermo Scientific Chemicals Supplier No. B2036514
Only null left

CAS: 1004-66-6 | C9H12O | 136.19 g/mol

2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1004-66-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00008380
InChI Key GFNZJAUVJCGWLW-UHFFFAOYSA-N
Synonym 2,6-dimethylanisole, 2-methoxy-m-xylene, benzene, 2-methoxy-1,3-dimethyl, 1,3-dimethyl-2-methoxybenzene, pubchem4115, 2,6-dimethyl anisole, acmc-1buuc, ksc493q8r, 2,6-dimethylphenyl methyl ether
PubChem CID 66088
IUPAC Name 2-methoxy-1,3-dimethylbenzene
SMILES COC1=C(C)C=CC=C1C

Specifications

Density 0.962
Boiling Point 181°C to 182°C
Quantity 25 g
Refractive Index 1.503
Beilstein 1858509
Solubility Information Insoluble in water.
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material 2,6-Dimethylanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-723-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only