missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline 99.0+%, TCI America™

Catalog No. p-7126360
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066925ML 25 mL
D0669100ML 100 mL
D0669500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D066925ML Supplier TCI America Supplier No. D066925ML
Only null left

Chemical Identifiers

CAS 87-62-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007747
InChI Key UFFBMTHBGFGIHF-UHFFFAOYSA-N
Synonym 2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
PubChem CID 6896
ChEBI CHEBI:28738
IUPAC Name 2,6-dimethylaniline
SMILES CC1=CC=CC(C)=C1N

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1711
Boiling Point 216°C
Formula Weight 121.18
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2,6-Dimethylaniline
EINECSNumber (3)-0129
RTECSNumber ZE9275000
TSCA Yes