missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline, 99%

Catalog No. p-115407
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115940050 5 g Glass bottle
AC115941000 100 g Glass bottle
AC115945000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC115940050 Supplier Thermo Scientific Chemicals Supplier No. 115940050
Only null left

CAS: 87-62-7 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-62-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007747
InChI Key UFFBMTHBGFGIHF-UHFFFAOYSA-N
Synonym 2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
PubChem CID 6896
ChEBI CHEBI:28738
IUPAC Name 2,6-dimethylaniline
SMILES CC1=CC=CC(C)=C1N

Specifications

Melting Point 10°C to 12°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9800g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5591 to 1.5611
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.98
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1107
Merck Index 15, 10282
Solubility Information Solubility in water: 7.5g/L (20 c). Other solubilities: very soluble in ether and ethanol,soluble in dimethyl sulfoxide,carbon tetrachlorid
Boiling Point 216°C
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2, 6-Dimethylaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-758-7
RTECSNumber ZE9275000
TSCA TSCA
Recommended Storage product darkens if exposed to light and air

RUO – Research Use Only