missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dimethylacetanilide, 97%

Catalog No. p-7028394
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2568714 25 g
AAB2568722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2568714 Supplier Thermo Scientific Chemicals Supplier No. B2568714

CAS: 2198-53-0 | C10H13NO | 163.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2198-53-0
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00008675
InChI Key NRPTXWYBRKRZES-UHFFFAOYSA-N
Synonym n-2,6-dimethylphenyl acetamide, 2',6'-dimethylacetanilide, 2,6-dimethylacetanilide, 2',6'-acetoxylidide, acetamide, n-2,6-dimethylphenyl, n-acetyl-2,6-xylidine, n-acetyl-2,6-dimethylaniline, acetanilide, 2',6'-dimethyl, unii-68gzi4j9fm, 68gzi4j9fm
PubChem CID 16616
IUPAC Name N-(2,6-dimethylphenyl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)C

Specifications

Melting Point 179°C to 183°C
Quantity 25 g
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 2',6'-Dimethylacetanilide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-596-8
RTECSNumber AN7470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only