missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-5-heptenal 85.0+%, TCI America™

Catalog No. D141325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D141325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D141325ML Supplier TCI America Supplier No. D141325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-72-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00006981
InChI Key YGFGZTXGYTUXBA-UHFFFAOYSA-N
Synonym 2,6-dimethyl-5-heptenal, melonal, 5-heptenal, 2,6-dimethyl, bergamal, 2,6-dimethyl-5-hepten-1-al, 2,6-dimethyl-5-heptanal, 2,6-dimethylhept-5-en-1-al, fema no. 2389, 2,6-dimethyl-2-hepten-7-al, 2,6-dimethyl-5-heptenal natural
PubChem CID 61016
IUPAC Name 2,6-dimethylhept-5-enal
SMILES CC(CCC=C(C)C)C=O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 mL
UN Number 1989
Formula Weight 140.23
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-5-heptenal
RTECSNumber MJ8797000
TSCA Yes