accessibility menu, dialog, popup

UserName

CAS RN 106-72-9

IUPAC Name: 2,6-dimethylhept-5-enal
Synonyms: dimethylheptenal dimethyl heptenal
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: YGFGZTXGYTUXBA-UHFFFAOYNA-N
SMILES: CC(CCC=C(C)C)C=O
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)