missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethyl-4-nitrophenol 98.0+%, TCI America™
Supplier: TCI America D253125G
Specifications
| 2,6-Dimethyl-4-nitrophenol | |
| 170°C | |
| MFCD00007339 | |
| 4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro | |
| CC1=CC(=CC(=C1O)C)[N+](=O)[O-] | |
| 167.164 | |
| 167.16 | |
| Crystalline Powder |
| 2423-71-4 | |
| C8H9NO3 | |
| 25 g | |
| FNORUNUDZNWQFF-UHFFFAOYSA-N | |
| 2,6-dimethyl-4-nitrophenol | |
| 17030 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2423-71-4 | |
| 167.164 | |
| FNORUNUDZNWQFF-UHFFFAOYSA-N | |
| 17030 | |
| CC1=CC(=CC(=C1O)C)[N+](=O)[O-] |
| C8H9NO3 | |
| MFCD00007339 | |
| 4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro | |
| 2,6-dimethyl-4-nitrophenol |
Safety and Handling
RTECSNumber : ZE8225000
TSCA : No