missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-nitrophenol 98.0+%, TCI America™

Catalog No. D25315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25315G 5 g
D253125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25315G Supplier TCI America Supplier No. D25315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2423-71-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007339
InChI Key FNORUNUDZNWQFF-UHFFFAOYSA-N
Synonym 4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro
PubChem CID 17030
IUPAC Name 2,6-dimethyl-4-nitrophenol
SMILES CC1=CC(=CC(=C1O)C)[N+](=O)[O-]

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-4-nitrophenol
RTECSNumber ZE8225000
TSCA No