missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-nitrophenol, 98%

CAS: 2423-71-4 | C8H9NO3 | 167.164 g/mol

$106.44 - $397.19

Chemical Identifiers

CAS 2423-71-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007339
InChI Key FNORUNUDZNWQFF-UHFFFAOYSA-N
PubChem CID 17030
IUPAC Name 2,6-dimethyl-4-nitrophenol
SMILES CC1=CC(=CC(=C1O)C)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1547206
View Documents
Thermo Scientific Chemicals
A1547206
5 g
Each for $106.44
Only null left
 
AAA1547214
View Documents
Thermo Scientific Chemicals
A1547214
25 g
Each for $397.19
Only null left
 
Description

Description

It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

2423-71-4
167.164
FNORUNUDZNWQFF-UHFFFAOYSA-N
2,6-dimethyl-4-nitrophenol
C8H9NO3
MFCD00007339
17030
CC1=CC(=CC(=C1O)C)[N+](=O)[O-]

Specifications

2423-71-4
C8H9NO3
5 g
4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro
FNORUNUDZNWQFF-UHFFFAOYSA-N
2,6-dimethyl-4-nitrophenol
17030
98%
∼168°C (decomposition)
MFCD00007339
1873275
Insoluble in water.
CC1=CC(=CC(=C1O)C)[N+](=O)[O-]
167.164
167.17
2,6-Dimethyl-4-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 219-353-9

RTECSNumber : ZE8225000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only