missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-nitroanisole, 99%

Catalog No. AAH2620403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2620403 1 g
AAH2620406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2620403 Supplier Thermo Scientific Chemicals Supplier No. H2620403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14804-39-8 | C9H11NO3 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14804-39-8
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00024537
InChI Key HSDNHFOJTRMGER-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-nitroanisole, m-xylene, 2-methoxy-5-nitro, benzene,2-methoxy-1,3-dimethyl-5-nitro, acmc-20ao84, m-xylene, 2-methoxy-5-nitro-,, 4-methoxy-3,5-dimethylnitrobenzene, 2,6-dimethyl-1-methoxy-4-nitrobenzene, 2-methoxy-1,3-di-methyl-5-nitrobenzene, benzene, 2-methoxy-1,3-dimethyl-5-nitro
PubChem CID 139835
IUPAC Name 2-methoxy-1,3-dimethyl-5-nitrobenzene
SMILES COC1=C(C)C=C(C=C1C)[N+]([O-])=O

Specifications

Melting Point 85°C to 92°C
Quantity 1 g
Formula Weight 181.19
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-4-nitroanisole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only