missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-3,5-heptanedione, 97+%

Catalog No. AA4200503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4200503 1 g
AA4200506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4200503 Supplier Thermo Scientific Chemicals Supplier No. 04200503
Only null left

Metal chelating agent; preparation of metal diketonates for MOCVD | CAS: 18362-64-6 | C9H16O2 | 156.225 g/mol

2,6-Dimethyl-3,5-heptanedione is a metal chelating agent and it is used in the preparation of metal diketonates for Metal-Organic Chemical Vapor Deposition (MOCVD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18362-64-6
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00015040
InChI Key CEGGECULKVTYMM-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3,5-heptanedione, diisobutyrylmethane, 3,5-heptanedione, 2,6-dimethyl, acmc-209ekw, 2,6-dimethyl-3,5-heptandione
PubChem CID 87597
IUPAC Name 2,6-dimethylheptane-3,5-dione
SMILES CC(C)C(=O)CC(=O)C(C)C

Specifications

Density 0.91 g/mL
Quantity 1 g
UN Number UN1224
Solubility Information Slightly Soluble in water. (3.9g/L) (25°C)
Formula Weight 156.22
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-3,5-heptanedione
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 242-234-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only