accessibility menu, dialog, popup

UserName

H3CCH3OOCH3CH3

CAS RN 18362-64-6

IUPAC Name: 2,6-dimethylheptane-3,5-dione
Synonyms: 2,6-dimethylheptane-3,5-dione
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: CEGGECULKVTYMM-UHFFFAOYSA-N
SMILES: CC(C)C(=O)CC(=O)C(C)C
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)