missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxyphenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 56°C |
| Boiling Point | 263°C |
| Quantity | 100 g |
| Formula Weight | 154.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,6-Dimethoxyphenol |
Safety and Handling
| EINECSNumber | (3)-4256&(3)-4415 |
| RTECSNumber | SL0900000 |
| TSCA | Yes |