missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyphenol 98.0+%, TCI America™

Catalog No. D0639500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0639500G 500 g
D063925G 25 g
D0639100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0639500G Supplier TCI America Supplier No. D0639500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-10-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064434
InChI Key KLIDCXVFHGNTTM-UHFFFAOYSA-N
Synonym syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol
PubChem CID 7041
ChEBI CHEBI:955
IUPAC Name 2,6-dimethoxyphenol
SMILES COC1=CC=CC(OC)=C1O

Specifications

Melting Point 56°C
Boiling Point 263°C
Quantity 500 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxyphenol
EINECSNumber (3)-4256&(3)-4415
RTECSNumber SL0900000
TSCA Yes