missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyphenol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015891
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015891 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015891 Supplier Merck Emd Millipore Supplier No. 53877100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,6-Dimethoxyphenol can find applications mainly as a substrate of laccases catalysis and also determine its activity in aqueous solution. It can also be used as a potential marker compound in particular for emission of smoke in the atmosphere from wood.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 91-10-1
Flash Point 140°C
Physical Form Neat
Linear Formula (CH3 O)2C6H3 OH
MDL Number MFCD00064434
Melting Point 50°C to 57°C; 53°C to 57°C
Boiling Point 261°C
Synonym Pyrogallol 1,3-dimethyl ether
Type Analytical Standard
Quantity 100 mg
Formula Weight 154.16
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less