missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxynaphthalene 98.0+%, TCI America™

Catalog No. p-7127771
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35195G 5 g
D351925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35195G Supplier TCI America Supplier No. D35195G
Only null left

Chemical Identifiers

CAS 5486-55-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00004063
InChI Key AHKDVDYNDXGFPP-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dimethoxy, 2,6-dimethoxy-naphthalene, 2,6-dimethoxy naphthalene, pubchem22734, naphthalene,6-dimethoxy, acmc-1ap1a, 2,6-dimethoxynaphthalene
PubChem CID 79627
IUPAC Name 2,6-dimethoxynaphthalene
SMILES COC1=CC2=CC=C(OC)C=C2C=C1

Specifications

Melting Point 154°C
Quantity 5 g
Formula Weight 188.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxynaphthalene