accessibility menu, dialog, popup

UserName

CH3OOCH3

CAS RN 5486-55-5

IUPAC Name: 2,6-dimethoxynaphthalene
Synonyms: 2,6-dimethoxynaphthalene
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: AHKDVDYNDXGFPP-UHFFFAOYSA-N
SMILES: COC1=CC2=CC=C(OC)C=C2C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)