missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxynaphthalene, 99%

Catalog No. AAH2836603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2836603 1 g
AAH2836606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2836603 Supplier Thermo Scientific Chemicals Supplier No. H2836603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5486-55-5 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5486-55-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00004063
InChI Key AHKDVDYNDXGFPP-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dimethoxy, 2,6-dimethoxy-naphthalene, 2,6-dimethoxy naphthalene, pubchem22734, naphthalene,6-dimethoxy, acmc-1ap1a, 2,6-dimethoxynaphthalene
PubChem CID 79627
IUPAC Name 2,6-dimethoxynaphthalene
SMILES COC1=CC2=CC=C(OC)C=C2C=C1

Specifications

Melting Point 152°C to 154°C
Quantity 1 g
Formula Weight 188.23
Percent Purity 99%
Chemical Name or Material 2,6-Dimethoxynaphthalene
EINECSNumber 226-816-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only