missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxybenzoic Acid 98.0+%, TCI America™
$115.57 - $115.57
Chemical Identifiers
| CAS | 1466-76-8 |
|---|---|
| Molecular Formula | C9H10O4 |
| Molecular Weight (g/mol) | 182.18 |
| MDL Number | MFCD00002437 |
| InChI Key | MBIZFBDREVRUHY-UHFFFAOYSA-N |
| Synonym | benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 |
| PubChem CID | 15109 |
| IUPAC Name | 2,6-dimethoxybenzoic acid |
| SMILES | COC1=CC=CC(OC)=C1C(O)=O |
Chemical Identifiers
| 1466-76-8 | |
| 182.18 | |
| MBIZFBDREVRUHY-UHFFFAOYSA-N | |
| 15109 | |
| COC1=CC=CC(OC)=C1C(O)=O |
| C9H10O4 | |
| MFCD00002437 | |
| benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 | |
| 2,6-dimethoxybenzoic acid |
Specifications
| 1466-76-8 | |
| White | |
| MFCD00002437 | |
| benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 | |
| COC1=CC=CC(OC)=C1C(O)=O | |
| 182.18 | |
| 182.18 | |
| Crystalline Powder |
| 185°C | |
| C9H10O4 | |
| 25 g | |
| MBIZFBDREVRUHY-UHFFFAOYSA-N | |
| 2,6-dimethoxybenzoic acid | |
| 15109 | |
| ≥98.0% (GC,T) | |
| 2,6-Dimethoxybenzoic Acid |
Safety and Handling
TSCA : Yes