Learn More
2,6-Dimethoxybenzoic acid, 99%
CAS: 1466-76-8 | C9H10O4 | 182.18 g/mol
Supplier: Thermo Scientific Chemicals A1441822
Description
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.
Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO, Ethyl Acetate and acetone.
Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 2,6-Dimethoxybenzoic acid | |
| ∼188°C (decomposition) | |
| C9H10O4 | |
| 100 g | |
| benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 | |
| MBIZFBDREVRUHY-UHFFFAOYSA-N | |
| 2,6-dimethoxybenzoic acid | |
| 15109 | |
| 99% |
| 1466-76-8 | |
| Odorless | |
| MFCD00002437 | |
| 2212190 | |
| Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO,Ethyl Acetate and acetone. | |
| COC1=CC=CC(OC)=C1C(O)=O | |
| 182.18 | |
| 182.18 |
Chemical Identifiers
| 1466-76-8 | |
| 182.18 | |
| MBIZFBDREVRUHY-UHFFFAOYSA-N | |
| 15109 | |
| COC1=CC=CC(OC)=C1C(O)=O |
| C9H10O4 | |
| MFCD00002437 | |
| benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043 | |
| 2,6-dimethoxybenzoic acid |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 215-985-4
RTECSNumber : DG8598725
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only