missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxybenzoic acid, 99%

Catalog No. AAA1441814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441814 25 g
AAA1441822 100 g
AAA1441836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1441814 Supplier Thermo Scientific Chemicals Supplier No. A1441814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1466-76-8 | C9H10O4 | 182.18 g/mol

It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO, Ethyl Acetate and acetone.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1466-76-8
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002437
InChI Key MBIZFBDREVRUHY-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043
PubChem CID 15109
IUPAC Name 2,6-dimethoxybenzoic acid
SMILES COC1=CC=CC(OC)=C1C(O)=O

Specifications

Melting Point ∼188°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 2212190
Solubility Information Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO,Ethyl Acetate and acetone.
Formula Weight 182.18
Percent Purity 99%
Chemical Name or Material 2,6-Dimethoxybenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-985-4
RTECSNumber DG8598725
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only