missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyaniline 98.0+%, TCI America™

Catalog No. D44801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44801G 1 g
D44805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44801G Supplier TCI America Supplier No. D44801G

Chemical Identifiers

CAS 2734-70-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00053934
InChI Key HQBJSEKQNRSDAZ-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dimethoxy, 2,6-dimethoxyphenylamine, 2,6-dimethoxybenzenamine, 2,6-dimethoxy-phenylamine, buttpark 33\04-69, 2 6-dimethoxyaniline, pubchem4419, 2,6-dimethoxy aniline, acmc-1cnwd, ksc497k1h
PubChem CID 95940
IUPAC Name 2,6-dimethoxyaniline
SMILES COC1=C(C(=CC=C1)OC)N

Specifications

Melting Point 76°C
Boiling Point 140°C
Quantity 1 g
Formula Weight 153.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxyaniline