missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyaniline, 97%

Catalog No. p-7029650
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2713903 1 g
AAH2713906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2713903 Supplier Thermo Scientific Chemicals Supplier No. H2713903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2734-70-5 | C8H11NO2 | 153.181 g/mol

2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2734-70-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00053934
InChI Key HQBJSEKQNRSDAZ-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dimethoxy, 2,6-dimethoxyphenylamine, 2,6-dimethoxybenzenamine, 2,6-dimethoxy-phenylamine, buttpark 33\04-69, 2 6-dimethoxyaniline, pubchem4419, 2,6-dimethoxy aniline, acmc-1cnwd, ksc497k1h
PubChem CID 95940
IUPAC Name 2,6-dimethoxyaniline
SMILES COC1=C(C(=CC=C1)OC)N

Specifications

Melting Point 76°C to 78°C
Odor Characteristic
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 153.18
Percent Purity 97%
Chemical Name or Material 2,6-Dimethoxyaniline
Hazard Category H302+H332-H311-H315-H319-H335-H373
Hazard Statement GHS H Statement
H311-H373-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only