missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxy-4-methylphenol 97.0+%, TCI America™

Catalog No. p-7127386
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25915G 5 g
D259125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25915G Supplier TCI America Supplier No. D25915G
Only null left

Chemical Identifiers

CAS 6638-05-7
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00017289
InChI Key ZFBNNSOJNZBLLS-UHFFFAOYSA-N
Synonym 4-methyl-2,6-dimethoxyphenol, 4-methylsyringol, 2,6-dimethoxy-p-cresol, phenol, 2,6-dimethoxy-4-methyl, p-cresol, 2,6-dimethoxy, 3,5-dimethoxy-4-hydroxytoluene, 4-hydroxy-3,5-dimethoxytoluene, unii-fjj5mye75w, fjj5mye75w, 2,6-dimethoxy-4-methyl-phenol
PubChem CID 240925
IUPAC Name 2,6-dimethoxy-4-methylphenol
SMILES CC1=CC(=C(C(=C1)OC)O)OC

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 168.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dimethoxy-4-methylphenol
TSCA No
Recommended Storage Refrigerator