Learn More
2,6-Diisopropylphenyl isocyanate, 94%
CAS: 28178-42-9 | C13H17NO | 203.29 g/mol
Supplier: Thermo Scientific Chemicals L1061306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,6-Diisopropylphenyl isocyanate | |
| 0.951 | |
| 107°C (224°F) | |
| 1.518 | |
| 5 g | |
| 2645721 | |
| 2,6-diisopropylphenyl isocyanate, 2,6-diisopropylphenylisocyanate, 2-isocyanato-1,3-diisopropylbenzene, benzene, 2-isocyanato-1,3-bis 1-methylethyl, isocyanic acid 2,6-diisopropylphenyl ester, isocyanic acid, 2,6-diisopropylphenyl ester, 1,3-bis 1-methylethyl-2-isocyanatobenzene, 2,6-bis 1-methylethyl phenyl isocyanate, 2-isocyanato-1,3-di propan-2-yl benzene, benzene, 1,3-bis 1-methylethyl-2-isocyanato | |
| CC(C)C1=CC=CC(C(C)C)=C1N=C=O | |
| 203.29 | |
| 203.29 |
| 28178-42-9 | |
| 116°C to 117°C (10 mmHg) | |
| C13H17NO | |
| MFCD00008882 | |
| UN2206 | |
| Moisture sensitive | |
| FEUFNKALUGDEMQ-UHFFFAOYSA-N | |
| 2-isocyanato-1,3-di(propan-2-yl)benzene | |
| 92259 | |
| 94% |
Chemical Identifiers
| 28178-42-9 | |
| 203.29 | |
| FEUFNKALUGDEMQ-UHFFFAOYSA-N | |
| 92259 | |
| CC(C)C1=CC=CC(C(C)C)=C1N=C=O |
| C13H17NO | |
| MFCD00008882 | |
| 2,6-diisopropylphenyl isocyanate, 2,6-diisopropylphenylisocyanate, 2-isocyanato-1,3-diisopropylbenzene, benzene, 2-isocyanato-1,3-bis 1-methylethyl, isocyanic acid 2,6-diisopropylphenyl ester, isocyanic acid, 2,6-diisopropylphenyl ester, 1,3-bis 1-methylethyl-2-isocyanatobenzene, 2,6-bis 1-methylethyl phenyl isocyanate, 2-isocyanato-1,3-di propan-2-yl benzene, benzene, 1,3-bis 1-methylethyl-2-isocyanato | |
| 2-isocyanato-1,3-di(propan-2-yl)benzene |
Safety and Handling
GHS H Statement
H330-H334-H318-H302-H315-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber : 248-885-4
RTECSNumber : CY8545000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only