missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylphenyl isocyanate, 94%

Catalog No. AAL1061303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1061303 1 g
AAL1061306 5 g
AAL1061314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1061303 Supplier Thermo Scientific Chemicals Supplier No. L1061303

CAS: 28178-42-9 | C13H17NO | 203.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28178-42-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.29
MDL Number MFCD00008882
InChI Key FEUFNKALUGDEMQ-UHFFFAOYSA-N
Synonym 2,6-diisopropylphenyl isocyanate, 2,6-diisopropylphenylisocyanate, 2-isocyanato-1,3-diisopropylbenzene, benzene, 2-isocyanato-1,3-bis 1-methylethyl, isocyanic acid 2,6-diisopropylphenyl ester, isocyanic acid, 2,6-diisopropylphenyl ester, 1,3-bis 1-methylethyl-2-isocyanatobenzene, 2,6-bis 1-methylethyl phenyl isocyanate, 2-isocyanato-1,3-di propan-2-yl benzene, benzene, 1,3-bis 1-methylethyl-2-isocyanato
PubChem CID 92259
IUPAC Name 2-isocyanato-1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=CC=CC(C(C)C)=C1N=C=O

Specifications

Density 0.951
Boiling Point 116°C to 117°C (10 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.518
Quantity 1 g
UN Number UN2206
Beilstein 2645721
Sensitivity Moisture sensitive
Formula Weight 203.29
Percent Purity 94%
Chemical Name or Material 2,6-Diisopropylphenyl isocyanate
Show More Show Less
Hazard Category H302+H312-H315-H319-H330-H335
Hazard Statement GHS H Statement
H330-H334-H318-H302-H315-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 248-885-4
RTECSNumber CY8545000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only