missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diiodopyridine 97.0+%, TCI America™

Catalog No. p-7128718
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52681G 1 g
D52685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52681G Supplier TCI America Supplier No. D52681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53710-17-1
Molecular Formula C5H3I2N
Molecular Weight (g/mol) 330.895
MDL Number MFCD07366728
InChI Key IHBAEQUKKOAEES-UHFFFAOYSA-N
PubChem CID 11427514
IUPAC Name 2,6-diiodopyridine
SMILES C1=CC(=NC(=C1)I)I

Specifications

Melting Point 188°C
Quantity 1 g
Formula Weight 330.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diiodopyridine