accessibility menu, dialog, popup

UserName

CAS RN 53710-17-1

IUPAC Name: 2,6-diiodopyridine
Synonyms: 2,6-diiodopyridine
Molecular Weight (g/mol): 330.9
Molecular Formula: C5H3I2N
InChi Key: IHBAEQUKKOAEES-UHFFFAOYSA-N
SMILES: IC1=CC=CC(I)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results